Electro-Luminescent Stopwatch, Sper Scientific
Supplier: Sper Scientific
810037C
14003-180EA 202.56 USD
14003-180
Electro-Luminescent Stopwatch, Sper Scientific
Timers Digital Stopwatches
Electro-Luminescence is a new technology which provides a clearer display than back-lit LCD while at the same time using less power.
  • A sonically welded case ensures excellent water resistance
  • Large ⁵/₈" high display for good visibility
  • Has a lap function, alarm clock and calendar
The result is a stopwatch easy to read in the dark which will provide about 4 hours of continuous illumination. The electro luminescent water resistant digital stopwatch is excellent for HPLC, TLC, electrophoresis, dark rooms, and nocturnal behavioral studies. Counts up to 24 hours with resolution of ¹/₁₀₀ second for the first 40 minutes and 1 second resolution thereafter.
Certifications: N.I.S.T. traceable certificate of compliance for 14003-180.
Delivery information: Button cell battery included.