AFFY75902
J75902-AE
101175-850EA 144.55 USD
101175-850
AFFY75902
7.5M solution of lithium chloride (LiCl) in high purity dH2O. Solution is 0.2μm filtered and dispensed into durable, square Nalgene bottles.
Contaminant Testing:
DNase (endo): Passes test
DNase (exo): Passes test
RNase: Passes test