Heavy Duty Industrial MultiMeter, Extech
Supplier: Extech
EX505 EX505-NIST
97049-964EA 308.14 USD
97049-964 10171-868
Heavy Duty Industrial MultiMeter, Extech
Electrical Meters Digital Multimeters
True RMS DMM with 4,000 count large LCD display, temperature function and double molded housing for waterproof protection
Safety Rating; CAT III-1000V, CAT IV-600V.
The EX505 also offers dual sensitivity frequency functions (electrical/electronic) and 1000V input protection on all functions. It comes complete with double molded test leads, magnetic hanging strap, Type K bead wire temp probe, carrying case and 9V battery. TRMS. Continuity/Diode.
Certifications: CE, UL, IP67.
Ordering information: 3-Year Warranty.