Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Defensin HNP-1 (human)

Supplier: BACHEM AMERICAS INC

H-9855.1000 H-9855.0500 H-9855.0100
H-9855.1000BAEA 1304.61 USD InStock
H-9855.1000BA H-9855.0500BA H-9855.0100BA
Defensin HNP-1 (human)
Proteins and Peptides
Sequence: H-Ala-Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys-OH
CAS-No.: 136661-76-2
Mol.weight: 3442.08
SumFormula: C₁₅₀H₂₂₂N₄₄O₃₈S₆
Bulk item no.: H-9855
Synonym(s): Human Neutrophil Peptide-1#HNP-1
EINECS no.: -
MDL no.: MFCD18910833

Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).

Defensins comprise a family of small, cationic peptides prevalent in neutrophil granulocytes of humans, rabbits, guinea pigs and rats as well as in rabbit lung macrophages. These peptides show sequence homology and contain 6 conserved disulfide-linked cysteines. In vitro, all defensins display a range of prominent antimicrobial activities against bacteria, fungi, and certain enveloped viruses. In addition, human and rabbit defensins exert potent cytotoxicity in vitro against various mammalian tumor cells. The defensins act on their targets by permeabilizing the plasma membranes.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty