Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Miraculin (1-20)

Supplier: BACHEM AMERICAS INC

H-9455.0005 H-9455.0001
H-9455.0005BAEA 752.88 USD InStock
H-9455.0005BA H-9455.0001BA
Miraculin (1-20)
Proteins and Peptides
Sequence: H-Asp-Ser-Ala-Pro-Asn-Pro-Val-Leu-Asp-Ile-Asp-Gly-Glu-Lys-Leu-Arg-Thr-Gly-Thr-Asn-OH
CAS-No.: 198694-37-0
Mol.weight: 2112.28
SumFormula: C₈₈H₁₄₆N₂₆O₃₄
Bulk item no.: H-9455
Synonym(s):
EINECS no.: -
MDL no.: MFCD00076869

The eicosapeptide DSAPNPVLDIDGEKLRTGTN corresponds to the N-terminal sequence of the taste-modifying 191 amino acid-containing protein miraculin produced by the berries of Richadella dulcifica.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty