(Des-Gly¹⁰,D-Tyr⁵,D-Ser(tBu)⁶,Pro-NHEt⁹)-LHRH Acetate, Bachem
4018587.0005 4018587.0001
H-8790.0001BAEA 420.99 USD
H-8790.0001BA H-8790.0005BA
(Des-Gly¹⁰,D-Tyr⁵,D-Ser(tBu)⁶,Pro-NHEt⁹)-LHRH Acetate, Bachem
LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
H-4224 corresponds to buserelin.
Impurity D of the Buserelin Ph. Eur. monograph.