Peptide WE-14
Supplier: Bachem
4015049.0001 4015049.0005
H-8680.0001BAEA 437.83 USD
H-8680.0001BA H-8680.0005BA
Peptide WE-14
Proteins and Peptides
WE-14, a chromogranin A-derived peptide, was isolated from a human ileal carcinoid tumor. Its sequence WSKMDQLAKELTAE is highly conserved in many mammalian species and flanked by typical processing sites. Such factors would indicate a potential physiological role for peptide WE-14.