Cyclo(-D-Glu-Ala-D-allo-Ile-Leu-D-Trp)
Supplier: Bachem
4018098.0005
H-8405.0005BAEA 1875.34 USD
H-8405.0005BA
Cyclo(-D-Glu-Ala-D-allo-Ile-Leu-D-Trp)
Proteins and Peptides
Endothelins are peptides with exceptional vasoconstrictor potency. They play an important role in intercellular communications.
BE18257B, an endothelin antagonist with high selectivity for the ET-A receptor subtype.