TRAP-6 (2-6) Trifluoroacetate, Bachem
4031304.0005 4031304.0025
H-8325.0005BAEA 247.64 USD
H-8325.0005BA H-8325.0025BA
TRAP-6 (2-6) Trifluoroacetate, Bachem
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
PAR-1 peptides are also called TRAP peptides (thrombin receptor-activating peptides).
FLLRN inhibits platelet aggregation induced by SFLLRN or thrombin.