Human IL-1alpha (223-250)
Supplier: Bachem
4030784.0500 4030784.1000
H-8290.1000BAEA 1239.48 USD
H-8290.1000BA H-8290.0500BA
Human IL-1alpha (223-250)
Interleukins are soluble proteins that act as local hormones. They possess powerful immunoregulatory properties.
This fragment is able to increase prostaglandin E₂ (PGE₂) production by fibroblasts in vitro without promoting the proliferation of thymocytes.