(D-Ala²,N-Me-Phe⁴,methionin(O)-ol⁵)-Enkephalin Trifluoroacetate, Bachem
4018038.0005
H-8270.0005BAEA 330.69 USD
H-8270.0005BA
(D-Ala²,N-Me-Phe⁴,methionin(O)-ol⁵)-Enkephalin Trifluoroacetate, Bachem
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
DAMME (FK 33-824) shows only slight preference for µ-receptors over δ-receptors and has no significant affinity for K-receptors. DAMME exerts a potent analgesic effect after systemic administration. It is one of the best-known analogs that has undergone fairly extensive clinical testing.