(D-Cys(tBu)²,Thr(tBu)⁶)-Leu-Enkephalin-Thr
4016945.0005
H-8170.0005BAEA 110.22 USD
H-8170.0005BA
(D-Cys(tBu)²,Thr(tBu)⁶)-Leu-Enkephalin-Thr
Proteins and Peptides
For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides'.
The tBu-modified hexapeptide BUBUC is the most potent δ-receptor-specific enkephalin analog.