Rat BNP-45 Trifluoroacetate
Supplier: Bachem
4030222.0500
H-8035.0500BAEA 620.06 USD
H-8035.0500BA
Rat BNP-45 Trifluoroacetate
Proteins and Peptides
BNP, originally isolated from porcine brain, belongs to the class of natriuretic peptides, having an amino acid sequence and a pharmacological activity spectrum (including diuretic-natriuretic, hypotensive, and vasorelaxant actions) remarkably similar to that of atrial natriuretic peptide (ANP). Meanwhile, BNP precursor molecules have also been shown to exist in human and rat cardiac tissue. Comparison of the precursor sequences demonstrates structural species differences between the mammalian BNPs, a finding which contrasts with the highly conserved ANP sequences.