H-Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg-OH Trifluoroacetate
Supplier: Bachem
4015583.0001 4015583.0005
H-8030.0005BAEA 1487.13 USD
H-8030.0005BA H-8030.0001BA
H-Cys-Ser-Arg-Ala-Arg-Lys-Gln-Ala-Ala-Ser-Ile-Lys-Val-Ala-Val-Ser-Ala-Asp-Arg-OH Trifluoroacetate
The 19-amino acid peptide CSRARKQAASIKVAVSADR, which corresponds to residues 2091-2108 of the laminin A chain (plus an N-terminal cysteine), is contained within the long arm of the A chain. It is capable of stimulating neuronal-like process formation to almost the same extent as laminin and competes with laminin for stimulation of neurite outgrowth.