(H-Lys-Ile-Leu-Asn-Asp-Leu-Ser-Ser-Asp-Ala-Pro-Gly-Val-Pro-Arg)₈-MAP

Supplier: BACHEM AMERICAS INC

4014741.0001
H-7655.0001BAEA 1075.42 USD
H-7655.0001BA
(H-Lys-Ile-Leu-Asn-Asp-Leu-Ser-Ser-Asp-Ala-Pro-Gly-Val-Pro-Arg)₈-MAP
Proteins and Peptides
Sequence: (H-Lys-Ile-Leu-Asn-Asp-Leu-Ser-Ser-Asp-Ala-Pro-Gly-Val-Pro-Arg)₈-MAP
CAS-No.:
Mol.weight: 13495.50
SumFormula: C₅₈₉H₁₀₀₂N₁₇₅O₁₈₅
Bulk item no.: H-7655
Synonym(s):
EINECS no.:
MDL no.: MFCD00239539

Multiple antigenic peptide (KILNDLSSDAPGVPR)₈-MAP, the sequence corresponds to rat CREB (136-150).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR