H-Gly-Arg-Gly-Asp-Ser-Cys-OH Trifluoroacetate
Supplier: Bachem
4012874.0005 4012874.0025
H-7255.0025BAEA 2230.07 USD
H-7255.0025BA H-7255.0005BA
H-Gly-Arg-Gly-Asp-Ser-Cys-OH Trifluoroacetate
GRGDSC can be linked covalently to carriers or surfaces functionalized with maleimido groups.