SAP-I
Supplier: Bachem
4013825.0001
H-6940.0001BAEA 110.22 USD
H-6940.0001BA
SAP-I
Proteins and Peptides
Sperm-activating peptide I (SAP I) isolated from sea urchin eggs, is capable of binding to and stimulating the respiration, motility, and cyclic nucleotide metabolism of sea urchin spermatozoa.