Splenopentin Acetate
Supplier: Bachem
4012856.0025
H-6930.0025BAEA 426.72 USD
H-6930.0025BA
Splenopentin Acetate
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
Splenopentin, RKEVY, is an immunomodulatory pentapeptide corresponding to the amino acid residues 32-36 of the splenic hormone splenin.