(D-Arg²)-Dermorphin (1-4) amide Trifluoroacetate, Bachem
4014718.0005
H-6755.0005BAEA 137.77 USD
H-6755.0005BA
(D-Arg²)-Dermorphin (1-4) amide Trifluoroacetate, Bachem
Proteins and Peptides
For other opioid peptides see, the product families ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.
YrFG-amide, which is more stable to enzymatic cleavage, is the most potent of the (D-Arg²)-dermorphin fragments. It produces significant antinociception in morphinetolerant mice.