Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

Abz-Ala-Gly-Leu-Ala-p-nitrobenzylamide

Supplier: BACHEM AMERICAS INC

H-6675.0025 H-6675.0100
H-6675.0025BAEA 550.89 USD InStock
H-6675.0025BA H-6675.0100BA
Abz-Ala-Gly-Leu-Ala-p-nitrobenzylamide
Proteins and Peptides
Sequence: Abz-Ala-Gly-Leu-Ala-p-nitrobenzylamide
CAS-No.: 100307-95-7
Mol.weight: 583.65
SumFormula: C₂₈H₃₇N₇O₇
Bulk item no.: H-6675
Synonym(s): AAGLAN
EINECS no.: -
MDL no.: MFCD00237463

A fluorogenic substrate for neutral metalloendopeptidases, e.g. Pseudomonas aeruginosa elastase, enkephalinase (NEP 24.11), and thermolysin. Abz-AGLA-Nba is also cleaved by atrolysin B and C.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty