Mouse;Rat Serpinin Trifluoroacetate
Supplier: Bachem
4062082.0500 4062082.1000
H-6672.0500BAEA 344.47 USD
H-6672.0500BA H-6672.1000BA
Mouse;Rat Serpinin Trifluoroacetate
Serpinin is a chromogranin A (CgA)-derived peptide. It has been identified to be a key molecule which induces dense-core secretory granules (DCG) biogenesis by up-regulating protease nexin-1 (PN-1) expression in an activity-dependent manner in endocrine cells.