Human Angiotensin (1-12) Trifluoroacetate
Supplier: Bachem
4061269.0005 4061269.0001
H-6608.0001BAEA 152.35 USD
H-6608.0001BA H-6608.0005BA
Human Angiotensin (1-12) Trifluoroacetate
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
Angiotensin 1-12 is cleaved in human cardiac tissue by chymase yielding angiotensin II.