(Hyp³,beta-(2-thienyl)-Ala⁵,Tyr(Me)⁸-psi(CH₂NH)Arg⁹)-Bradykinin Trifluoroacetate, Bachem
Supplier: BACHEM AMERICAS INC
4057654.0001
H-6578.0001BAEA 457.2 USD
H-6578.0001BA
(Hyp³,beta-(2-thienyl)-Ala⁵,Tyr(Me)⁸-psi(CH₂NH)Arg⁹)-Bradykinin Trifluoroacetate, Bachem
Proteins and Peptides
H-6578 corresponds to the selective bradykinin B₂ receptor agonist labradimil.
Labradimil , Arg-Pro-Hyp-Gly-Thi-Ser-Pro-Tyr(Me)-psi(CH₂NH)-Arg, is a selective bradykinin B₂ receptor agonist with a longer plasma half-life than bradykinin. Labradimil transiently increases the permeability of the blood-brain barrier (BBB) to facilitate delivery of drugs to the CNS.