Alpha-Defensin 6 Trifluoroacetate
Supplier: Bachem
4059148.0500 4059148.0100
H-6566.0500BAEA 1347.3 USD
H-6566.0500BA H-6566.0100BA
Alpha-Defensin 6 Trifluoroacetate
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).
Defensins comprise a family of small, cationic peptides prevalent in neutrophil granulocytes of humans, rabbits, guinea pigs and rats as well as in rabbit lung macrophages. These peptides show sequence homology and contain 6 conserved disulfide-linked cysteines. In vitro, all defensins display a range of prominent antimicrobial activities against bacteria, fungi, and certain enveloped viruses. In addition, human and rabbit defensins exert potent cytotoxicity in vitro against various mammalian tumor cells. The defensins act on their targets by permeabilizing the plasma membranes.
Cysteine-rich cationic peptide with antiviral activity against HSV infection and a role in N. gonorrhoeae-mediated HIV enhancement. In contrast to the other members of the ƒ-defensin family it shows very low antimicrobial activity against Gram-negative and Gram-positive bacteria. As a biomarker of colon adenocarcinoma it may have utility in colon cancer screening and diagnosis.