Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

(Des-Thr⁵)-Glucagon

Supplier: BACHEM AMERICAS INC

H-6156.1000 H-6156.0500
H-6156.1000BAEA 537.12 USD InStock
H-6156.1000BA H-6156.0500BA
(Des-Thr⁵)-Glucagon
Proteins and Peptides
Sequence: H-His-Ser-Gln-Gly-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-OH
CAS-No.: 1802078-27-8
Mol.weight: 3381.69
SumFormula: C₁₄₉H₂₁₈N₄₂O₄₇S
Bulk item no.: H-6156
Synonym(s):
EINECS no.:
MDL no.: MFCD18782363

For the long-acting GLP-1 analog liraglutide see H-6724.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty