Salusin-α (human)

H-6034.1000 H-6034.0500
H-6034.1000BAEA 681.91 USD InStock
H-6034.1000BA H-6034.0500BA
Salusin-α (human)
Proteins and Peptides
Sequence: H-Ser-Gly-Ala-Leu-Pro-Pro-Ala-Pro-Ala-Ala-Pro-Arg-Pro-Ala-Leu-Arg-Ala-Gln-Arg-Ala-Gly-Pro-Ala-Gly-Pro-Gly-Ala-Lys-NH₂
CAS-No.: 624735-22-4
Mol.weight: 2603.03
SumFormula: C₁₁₄H₁₉₂N₄₀O₃₀
Bulk item no.: H-6034
Synonym(s):
EINECS no.: -
MDL no.:

Salusin-α and salusin-β are translated from an alternatively spliced mRNA of TOR2A, a gene encoding a protein of the torsion dystonia family. Salusins are recently identified regulators of hemodynamics and mitogenesis with no sequence homology to previously identified peptides. They are expressed and synthesized within human tissues and are present in human body fluids, suggesting that salusin-α and salusin-β are endocrine and / or paracrine factors.

Salusin-α, a bioactive peptide with mild hypotensive and bradycardic effects, could serve as biomarker for atherosclerotic coronary heart disease.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR