Pig Cytochrome C (88-104) Trifluoroacetate
Supplier: Bachem
4044055.0001
H-6016.0001BAEA 254.38 USD
H-6016.0001BA
Pig Cytochrome C (88-104) Trifluoroacetate
Proteins and Peptides
See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.
KAERADLIAYLKQATAK, an antigenic sequence used to study the T-cell response.