(Phe¹,Ser²)-TRAP-6 Trifluoroacetate, Bachem
4043420.0005
H-5996.0005BAEA 304.53 USD
H-5996.0005BA
(Phe¹,Ser²)-TRAP-6 Trifluoroacetate, Bachem
Proteins and Peptides
PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.
PAR-1 peptides are also called TRAP peptides (thrombin receptor-activating peptides).
The scrambled peptide (Phe¹,Ser²)-TRAP-6 (FSLLRN) may serve as an inactive control for TRAP-6, SFLLRN (H-8365).