Mouse Orphan GPCR SP9155 Agonist P550 Trifluoroacetate
Supplier: Bachem
4043083.1000 4043083.0500
H-5986.1000BAEA 842 USD
H-5986.1000BA H-5986.0500BA
Mouse Orphan GPCR SP9155 Agonist P550 Trifluoroacetate
P550 (mouse) is the mouse homolog of P518 (human). It has been shown to exhibit similar agonistic activity towards the human orphan G-protein-coupled receptor SP9155 as P518 (human) (EC₅₀ = 6 nM).