Influenza A Matrix Protein M1 (58-66) Acetate
Supplier: Bachem
4033339.0005
H-5874.0005BAEA 551.16 USD
H-5874.0005BA
Influenza A Matrix Protein M1 (58-66) Acetate
Proteins and Peptides
This influenza matrix peptide (58-66) was shown to be optimal for binding to HLA-2 and for presentation to cytotoxic T lymphocytes.