PYX-1 Trifluoroacetate
Supplier: Bachem
4039889.1000 4039889.0500
H-5786.1000BAEA 1002.68 USD
H-5786.1000BA H-5786.0500BA
PYX-1 Trifluoroacetate
Potent neuropeptide Y receptor antagonist. It inhibited neuropeptide Y action in releasing intracellular calcium in human erythroleukemia cells at a range of 10⁻⁶ to 10⁻⁷ M.