Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

β-Endorphin (equine)

Supplier: BACHEM AMERICAS INC

H-5636.0005 H-5636.0001
H-5636.0005BAEA 2101.94 USD InStock
H-5636.0005BA H-5636.0001BA
β-Endorphin (equine)
Proteins and Peptides
Sequence: H-Tyr-Gly-Gly-Phe-Met-Ser-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr-Leu-Phe-Lys-Asn-Ala-Ile-Ile-Lys-Asn-Ala-His-Lys-Lys-Gly-Gln-OH
CAS-No.: 79495-86-6
Mol.weight: 3423.99
SumFormula: C₁₅₄H₂₄₈N₄₂O₄₄S
Bulk item no.: H-5636
Synonym(s):
EINECS no.: -
MDL no.: MFCD00151074

For other opioid peptides see, the product families ‘Deltorphins and Dermorphins’, ‘Dynorphin, Analogs and Sequences, ‘Endorphins, MPF, Neoendorphins’, and ‘Opioid Peptides’.

β-Endorphin (equine) exhibited three times higher receptor binding activity in rat membrane preparations and a 1.6 times more potent analgesic effect in the mouse tail-flick assay than the human hormone.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty