Acetyl-(D-Arg²)-GRF (1-29) amide (human)

Supplier: BACHEM AMERICAS INC

4030691.0500 4030691.1000
H-5560.0500BAEA 263.79 USD InStock
H-5560.0500BA H-5560.1000BA
Acetyl-(D-Arg²)-GRF (1-29) amide (human)
Proteins and Peptides
Sequence: Ac-Tyr-D-Arg-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH₂
CAS-No.: 93942-91-7
Mol.weight: 3485.08
SumFormula: C₁₅₄H₂₅₅N₄₇O₄₃S
Bulk item no.: H-5560
Synonym(s): Acetyl-(D-Arg²)-Sermorelin#Ac-[DArg2] GHRF(1-29), amide, human
EINECS no.: -
MDL no.: MFCD18782292

For sermorelin see H-3705.

GRF antagonist.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR