H-Val-Leu-Ser-Glu-Gly-OH
Supplier: Bachem
4000376.0050
H-5285.0050BAEA 304.53 USD
H-5285.0050BA
H-Val-Leu-Ser-Glu-Gly-OH
Proteins and Peptides
VLSEG, a sequence from the N-terminal region of rat myoglobin.