Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Acetyl-(Asn³⁰,Tyr³²)-Calcitonin (8-32) (salmon I)

Supplier: BACHEM AMERICAS INC

H-4922.0500 H-4922.1000
H-4922.0500BAEA 537.12 USD InStock
H-4922.0500BA H-4922.1000BA
Acetyl-(Asn³⁰,Tyr³²)-Calcitonin (8-32) (salmon I)
Proteins and Peptides
Sequence: Ac-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asn-Thr-Gly-Ser-Asn-Thr-Tyr-NH₂
CAS-No.: 151804-77-2
Mol.weight: 2890.25
SumFormula: C₁₂₇H₂₀₅N₃₇O₄₀
Bulk item no.: H-4922
Synonym(s): AC187
EINECS no.: -
MDL no.: MFCD03453640

For Elcatonin see H-2214.

AC187 is a potent and selective antagonist of amylin-induced hyperglycaemia and hyperlactaemia. It has been used to demonstrate that amylin receptors mediate the anorectic action of salmon calcitonin.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty