Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ovalbumin (257-264) (chicken)

Supplier: BACHEM AMERICAS INC

H-4866.0001 H-4866.0005
H-4866.0001BAEA 125.32 USD InStock
H-4866.0001BA H-4866.0005BA
Ovalbumin (257-264) (chicken)
Proteins and Peptides
Sequence: H-Ser-Ile-Ile-Asn-Phe-Glu-Lys-Leu-OH
CAS-No.: 138831-86-4
Mol.weight: 963.14
SumFormula: C₄₅H₇₄N₁₀O₁₃
Bulk item no.: H-4866
Synonym(s): OVA (257-264) (chicken)#OVA peptide (257-264)
EINECS no.: -
MDL no.: MFCD18452854

SIINFEKL, an ovalbumin-derived peptide, is known to arise from natural processing of ovalbumin H-2b mouse cells. Cytotoxic T lymphocytes (CTL) from the ovalbumin-hsp70-immunized mice and a highly effective CTL clone (4G3) raised against ovalbumin-expressing EL4 tumor cells (EG7-OVA) were equally effective in terms of the concentration of this compound required for half-maximal lysis in a CTL assay.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty