H-Lys-Lys-Lys-OH

Supplier: BACHEM AMERICAS INC

4002495.0250 H-4175.0050
H-4175.0250BAEA 669.6 USD InStock
H-4175.0250BA H-4175.0050BA
H-Lys-Lys-Lys-OH
Proteins and Peptides
Sequence: H-Lys-Lys-Lys-OH
CAS-No.: 13184-14-0
Mol.weight: 402.54
SumFormula: C₁₈H₃₈N₆O₄
Bulk item no.: H-4175
Synonym(s):
EINECS no.: -
MDL no.: MFCD00238054

Trilysine.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR