(D-Ala⁶)-LHRH

Supplier: BACHEM AMERICAS INC

4009745.0005 4009745.0025
H-4020.0005BAEA 286.46 USD
H-4020.0005BA H-4020.0025BA
(D-Ala⁶)-LHRH
Proteins and Peptides
Sequence: Pyr-His-Trp-Ser-Tyr-D-Ala-Leu-Arg-Pro-Gly-NH₂
CAS-No.: 51230-19-4
Mol.weight: 1196.33
SumFormula: C₅₆H₇₇N₁₇O₁₃
Bulk item no.: H-4020
Synonym(s): (D-Ala⁶)-GnRH#(D-Ala⁶)-Gonadorelin#[DAla6] LH-RH
EINECS no.: -
MDL no.: MFCD00133503

LHRH is also called Gonadotropin-Releasing Hormone (GnRH) or Luteinizing Hormone-Releasing Factor (LRF).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR