H-Leu-Leu-Tyr-OH

Supplier: BACHEM AMERICAS INC

4009556.0250 4009556.0050
H-3940.0250BAEA 716.16 USD InStock
H-3940.0250BA H-3940.0050BA
H-Leu-Leu-Tyr-OH
Proteins and Peptides
Sequence: H-Leu-Leu-Tyr-OH
CAS-No.: 20368-24-5
Mol.weight: 407.51
SumFormula: C₂₁H₃₃N₃O₅
Bulk item no.: H-3940
Synonym(s):
EINECS no.: -
MDL no.: MFCD00057825

See also the product families 'Adjuvant Peptides' and 'Thymosins and Fragments'.

Immunostimulating tripeptide from human and cow casein. LLY corresponds to bovine β-casein 191-193. See also H-Gly-Leu-Phe-OH.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR