Kentsin
Supplier: Bachem
4003563.0025
H-3840.0025BAEA 137.77 USD
H-3840.0025BA
Kentsin
Proteins and Peptides
Kentsin (contraceptive tetrapeptide) was first isolated from the oviductal lumen of progravid female hamsters. Kentsin shows opioid-like activity without binding to opioid receptors.