Caerulein

Supplier: BACHEM AMERICAS INC

4030451.0005 4030451.0001
H-3220.0005BAEA 904.71 USD InStock
H-3220.0005BA H-3220.0001BA
Caerulein
Proteins and Peptides
Sequence: Pyr-Gln-Asp-Tyr(SO₃H)-Thr-Gly-Trp-Met-Asp-Phe-NH₂
CAS-No.: 17650-98-5
Mol.weight: 1352.42
SumFormula: C₅₈H₇₃N₁₃O₂₁S₂
Bulk item no.: H-3220
Synonym(s): Ceruletide#Cerulein
EINECS no.: -
MDL no.: MFCD00076478

This sulfated peptide isolated first from the skin of the Australian frog Litoria caerulea is a more potent secretagogue than its analog CCK-8. Ceruletide is applied in the diagnosis of pancreatic function and, in high dosage, for the induction of pancreatitis in experimental animals.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR