Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TRAP-6 amide

Supplier: BACHEM AMERICAS INC

H-2936.0005 H-2936.0025
H-2936.0005BAEA 87.82 USD InStock
H-2936.0005BA H-2936.0025BA
TRAP-6 amide
Proteins and Peptides
Sequence: H-Ser-Phe-Leu-Leu-Arg-Asn-NH₂
CAS-No.: 141923-40-2
Mol.weight: 747.90
SumFormula: C₃₄H₅₇N₁₁O₈
Bulk item no.: H-2936
Synonym(s): SFLLRNamide#Thrombin Receptor Activator Peptide 6 amide#PAR-1 (1-6) amide (human)#Proteinase Activated Receptor 1 (1-6) amide (human)#Thrombin Receptor (1-6) amide (human)#Coagulation Factor II Receptor (1-6) amide (human)#SFLLRN-NH2 Selective Protease-Activated Receptor 1 (PAR1) Agonist
EINECS no.: -
MDL no.: MFCD18782166

PAR peptides (TRAP peptides and thrombin receptor-like peptides) activate the proteinase-activated receptors PAR-1 to PAR-4.

PAR-1 peptides are also called TRAP peptides (thrombin receptor-activating peptides).

SFLLRN-amide, thrombin receptor agonist peptide.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty