Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.

(Met⁵,Pro⁶,D-Phe⁷,D-Trp⁹,Phe¹⁰)-α-MSH (5-13)

Supplier: BACHEM AMERICAS INC

H-2716.0005 H-2716.0001
H-2716.0005BAEA 383.14 USD InStock
H-2716.0005BA H-2716.0001BA
(Met⁵,Pro⁶,D-Phe⁷,D-Trp⁹,Phe¹⁰)-α-MSH (5-13)
Proteins and Peptides
Sequence: H-Met-Pro-D-Phe-Arg-D-Trp-Phe-Lys-Pro-Val-NH₂
CAS-No.: 158563-45-2
Mol.weight: 1206.52
SumFormula: C₆₁H₈₇N₁₅O₉S
Bulk item no.: H-2716
Synonym(s):
EINECS no.: -
MDL no.: MFCD00672435

Potent α-MSH antagonist (IC₅₀ = 11 ± 7 nM).

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty