H-Tyr-Ile-Tyr-Gly-Ser-Phe-Lys-OH
Supplier: Bachem
4031166.0005 4031166.0001
H-2686.0001BAEA 110.22 USD
H-2686.0001BA H-2686.0005BA
H-Tyr-Ile-Tyr-Gly-Ser-Phe-Lys-OH
The octapeptide YIYGSFK selected from a random combinatorial peptide library is a much more specific and efficient substrate for the Src-family protein tyrosine kinase than cdc2 (6-20), a peptide derived from 34p cdc2. Its Km value for p60 c-src (55 µM) is about 6.4-fold lower than that of the cdc2 (6-20) substrate.