Antistasin-Related Peptide
4026402.0001
H-2674.0001BAEA 304.53 USD
H-2674.0001BA
Antistasin-Related Peptide
Anticoagulant and antithrombotic peptides. Please see also the product family 'Hirudin Fragments and Analogs', the inhibitors N-1065, N-1215, N-1260, N-1565, and N-1970, and the anticoagulant anisindione, Q-1085.
Antistasin, isolated from the Mexican leech Hementeria officinalis, is a 119 amino acid protein which is a selective and potent inhibitor of coagulation factor Xa. (D-Arg³²)-Antistasin (32-38) is the shortest peptide displaying anticoagulant activity in human plasma. This peptide might be a model structure for the development of low molecular weight anticoagulants.