Amyloid beta/A4 Protein Precursor₇₇₀ (394-410) Trifluoroacetate
Supplier: Bachem
4026205.0001 4026205.0005
H-2594.0005BAEA 871.91 USD
H-2594.0005BA H-2594.0001BA
Amyloid beta/A4 Protein Precursor₇₇₀ (394-410) Trifluoroacetate
See also the product family 'Secretase Inhibitors and Substrates'.comprising products related to β- and γ-secretase.
The 17-mer peptide AKERLEAKHRERMSQVM is capable of reducing the neurologic damage in vivo in a model of central nervous system ischemia, possibly through its effect on synaptic plasticity.