Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodefrin

Supplier: BACHEM AMERICAS INC
H-2566.0005 H-2566.0001
Sequence: H-Ser-Ile-Pro-Ser-Lys-Asp-Ala-Leu-Leu-Lys-OH
CAS-No.: 161928-96-7
Mol.weight: 1071.28
SumFormula: C₄₈H₈₆N₁₂O₁₅
Bulk item no.: H-2566
Synonym(s):
EINECS no.: -
MDL no.: MFCD00678073

The amphibian skin is a rich source of antimicrobial peptides, please see the same-named product family Other amphibian peptides as bombesin, caerulein, deltorphin, dermorphin, pseudin, sauvagine, and xenin show considerable pharmacological activity.

Sodefrin has first been isolated from the abdominal gland of the cloaca of the male red-bellied newt, Cynops pyrrhogaster. It showed a species specific female-attracting activity. It is the first amphibian pheromone to be identified and the first peptide pheromone identified in a vertebrate.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1 mg H-2566.0001 H-2566.0001BA Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

5 mg H-2566.0005 H-2566.0005BA Each (5mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Sodefrin
CAS No. 161928-96-7
Molecular Weight 1071.28
Molecular Formula C₄₈H₈₆N₁₂O₁₅
Storage Conditions -20 ± 5 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping