Mouse SPARC (119-122) Acetate
Supplier: Bachem
4026105.0050 4026105.0250
H-2472.0250BAEA 892.77 USD
H-2472.0250BA H-2472.0050BA
Mouse SPARC (119-122) Acetate
SPARC is a transiently expressed extracellular matrix-binding protein that alters cell shape and regulates endothelial cell proliferation in vitro. KGHK, a copper-binding peptide fragment, has been shown to be a potent stimulator of angiogenesis. Specific degradation of SPARC by proteases of the extracellular fluid may release this bioactive tetrapeptide. Acetylated KGHK is used as cosmetic ingredient.