H-Ala-D-Glu(Lys-D-Ala-D-Ala-OH)-OH, Bachem
Supplier: Bachem
4025962.0025 4025962.0005
H-2416.0025BAEA 993.47 USD
H-2416.0025BA H-2416.0005BA
H-Ala-D-Glu(Lys-D-Ala-D-Ala-OH)-OH, Bachem
Proteins and Peptides
See also F-3260.
A murein peptide sequence. Substrate for DD-carboxypeptidase (muramoylpentapeptide carboxypeptidase, D-alanine carboxypeptidase).