(d(CH₂)₅¹,D-Ile²,Ile⁴,Arg⁸)-Vasopressin
4031345.0001
H-2404.0001BAEA 275.57 USD
H-2404.0001BA
(d(CH₂)₅¹,D-Ile²,Ile⁴,Arg⁸)-Vasopressin
Proteins and Peptides
For desmopressin see H-7675.
Potent and selective antagonist of the antidiuretic response to AVP ((Arg⁸)-Vasopressin) with an antivasopressor/antidiuretic effective dose ratio of 39.